missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15420-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize both RAB2A and RAB2B.
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.Specifications
| RAB2 | |
| Polyclonal | |
| Unconjugated | |
| RAB2A | |
| RAB2, RAB2A, Ras related protein Rab 2A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305853, 5862, 59021, 65158, 76338, 84932 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05712, P53994, P59279, P61019, Q8WUD1 | |
| Rab2a, RAB2B | |
| RAB2 Fusion Protein Ag7708 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction