missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118228
This item is not returnable.
View return policy
Description
RAB1A Polyclonal Antibody for Western Blot, IP
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
Specifications
| RAB1A | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P62820 | |
| RAB1A | |
| E. coli-derived Human RAB1A fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| RAB1A | |
| Ac2-048; DKFZP564B163; Gtbp; GTP binding protein Rab1a; LOW QUALITY PROTEIN: ras-related protein Rab-1A; mKIAA3012; Rab GTPase YPT1 homolog; RAB1; Rab-1; RAB1, member RAS oncogene family; Rab1A; RAB1A, member RAS oncogene family; RAB1A-like protein; Rab1r; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1; YPT1-related protein | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction