missing translation for 'onlineSavingsMsg'
Learn More

Rab1A Mouse anti-Human, Mouse, Clone: 7H4, Novus Biologicals™

Product Code. 18338592 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338592 25 μg 25µg
18365243 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18338592 Supplier Novus Biologicals Supplier No. NBP31104225UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rab1A Monoclonal specifically detects Rab1A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab1A
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 7H4
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation
Formulation PBS (pH 7.4), 50% Glycerol
Gene Alias DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A
Host Species Mouse
Immunogen Full length recombinant Rab1A protein (NP_004152.1)
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5861
Target Species Human, Mouse
Content And Storage Store at -20°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.