missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Rab1A Monoclonal specifically detects Rab1A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
| Antigen | Rab1A |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 4G10 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunoprecipitation |
| Formulation | PBS (pH 7.4), 50% Glycerol |
| Gene Alias | DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?