missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104435
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB19.
RAB19 gene ontology annotations related to this gene include endoplasmic reticulum to Golgi vesicle-mediated transport; intracellular protein transport; Rab protein signal transduction.
Specifications
| RAB19 | |
| Polyclonal | |
| Unconjugated | |
| RAB19 | |
| GTP-binding protein RAB19B; hypothetical protein LOC506911; Rab19; RAB19, member RAS oncogene family; RAB19B; Ras-related protein Rab-19; ras-related protein Rab-19-like; zmp:0000001111 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19331, 401409, 500088 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A4D1S5, P35294, Q5M7U5 | |
| RAB19 | |
| A synthesized peptide derived from human RAB19(Accession A4D1S5), corresponding to amino acid residues V140-E190. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction