missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11304-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of a family of Ras-related small GTPases that regulate membrane trafficking in organelles and transport vesicles. Knockdown studies is zebrafish suggest that this protein may have a role in eye and brain development. Mutations in this gene are associated with Warburg micro syndrome type 3. Alternatively spliced transcript variants have been found for this gene.Specifications
| Rab18 | |
| Polyclonal | |
| Unconjugated | |
| RAB18 | |
| RAB18, RAB18LI1, Ras related protein Rab 18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19330, 22931, 307039 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35293, Q5EB77, Q9NP72 | |
| Rab18 | |
| Rab18 Fusion Protein Ag1841 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction