Learn More
Rab18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11304-1-AP-150UL
 View more versions of this product
Product Code. 16878283
  • £349.00 / 150µL
Description
The protein encoded by this gene is a member of a family of Ras-related small GTPases that regulate membrane trafficking in organelles and transport vesicles. Knockdown studies is zebrafish suggest that this protein may have a role in eye and brain development. Mutations in this gene are associated with Warburg micro syndrome type 3. Alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
Rab18
Polyclonal
Unconjugated
RAB18
RAB18, RAB18LI1, Ras related protein Rab 18
Rabbit
Antigen Affinity Chromatography
RUO
19330, 22931, 307039
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P35293, Q5EB77, Q9NP72
Rab18
Rab18 Fusion Protein Ag1841
150 μL
Primary
Human, Mouse, Rat