missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB18 Antibody (1G11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43021
This item is not returnable.
View return policy
Description
RAB18 Monoclonal antibody specifically detects RAB18 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| RAB18 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1G11 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| RAB18 small GTPase, RAB18, member RAS oncogene family, RAB18LI1, ras-related protein Rab-18 | |
| RAB18, member RAS oncogene family - (Uniprot# Q9NP72) | |
| 100 μg | |
| Signal Transduction | |
| 22931 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction