missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115816
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB17.
The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase.
Specifications
| RAB17 | |
| Polyclonal | |
| Unconjugated | |
| Rab17 | |
| AW413472; Rab17; RAB17, member RAS oncogene family; ras-related protein Rab-17; RGD1562478 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19329, 503269, 64284 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P35292, Q9H0T7 | |
| Rab17 | |
| A synthesized peptide derived from human RAB17(Accession Q9H0T7), corresponding to amino acid residues S82-N132. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction