missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab17 Antibody (OTI5E2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73765CL1
This item is not returnable.
View return policy
Description
Rab17 Monoclonal antibody specifically detects Rab17 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Rab17 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ12538, RAB17, member RAS oncogene family, ras-related protein Rab-17 | |
| Full length human recombinant protein of human RAB17(NP_071894) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 64284 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI5E2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction