missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB14 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93227-0.02ml
This item is not returnable.
View return policy
Description
RAB14 Polyclonal antibody specifically detects RAB14 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RAB14 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| bA165P4.3 (member RAS oncogene family), F protein-binding protein 1, FBP, RAB-14, RAB14, member RAS oncogene family, ras-related protein Rab-14, small GTP binding protein RAB14 | |
| A synthetic peptide corresponding to a sequence within amino acids 39-140 of human RAB14 (NP_057406.2). DCPHTIGVEFGTRIIEVSGQKIKLQIWDTAGQERFRAVTRSYYRGAAGALMVYDITRRSTYNHLSSWLTDARNLTNPNTVIILIGNKADLEAQRDVTYEEAK | |
| 0.02 mL | |
| Signal Transduction | |
| 51552 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction