missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568299
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
RAB13 Polyclonal antibody specifically detects RAB13 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| RAB13 | |
| Polyclonal | |
| Unconjugated | |
| RAB13 | |
| 0610007N03Rik; B230212B15Rik; Cell growth-inhibiting gene 4 protein; GIG4; growth-inhibiting gene 4 protein; Rab13; RAB13, member RAS oncogene family; RAS-associated protein RAB13; ras-related protein Rab-13 | |
| Rabbit | |
| affinity chromatography | |
| RUO | |
| 5872, 68328, 81756 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35286, P51153, Q9DD03 | |
| RAB13 | |
| Synthesized peptide derived from human RAB13. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction