missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118915
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB13.
RAB13 can participate in polarized transport, in the assembly and/or the activity of tight junctions.
Specifications
| RAB13 | |
| Polyclonal | |
| Unconjugated | |
| RAB13 | |
| 0610007N03Rik; B230212B15Rik; Cell growth-inhibiting gene 4 protein; GIG4; growth-inhibiting gene 4 protein; Rab13; RAB13, member RAS oncogene family; RAS-associated protein RAB13; ras-related protein Rab-13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5872, 68328, 81756 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35286, P51153, Q9DD03 | |
| RAB13 | |
| A synthesized peptide derived from human RAB13(Accession P51153), corresponding to amino acid residues E68-L118. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction