missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB13 Monoclonal Antibody (8E8E2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531878
This item is not returnable.
View return policy
Description
MA5-31878 has been tested in indirect ELISA.
RAB13 can participate in polarized transport, in the assembly and/or the activity of tight junctions.
Specifications
| RAB13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P51153 | |
| RAB13 | |
| Purified recombinant fragment of human Rab13 (AA: 66-200) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 8E8E2 | |
| Unconjugated | |
| RAB13 | |
| 0610007N03Rik; B230212B15Rik; Cell growth-inhibiting gene 4 protein; GIG4; growth-inhibiting gene 4 protein; Rab13; RAB13, member RAS oncogene family; RAS-associated protein RAB13; ras-related protein Rab-13 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction