missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB11FIP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557642
This item is not returnable.
View return policy
Description
Immunogen sequence: SPHPVKPLSA APVEGSPDRK QSRSSLSIAL SSGLEKLKTV TSGSIQPVTQ APQAGQMVDT KRLKDSAVLD QSAKYYHLTH DELISLLLQR ERELSQRDEH VQEL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
Rab effector involved in protein trafficking from apical recycling endosomes to the apical plasma membrane. Involved in insulin granule exocytosis. May regulate V-ATPase intracellular transport in response to extracellular acidosis.
Specifications
| RAB11FIP5 | |
| Polyclonal | |
| Unconjugated | |
| RAB11FIP5 | |
| 9130206P09Rik; C75969; D6Ertd32e; GAF1; Gaf-1; Gamma-SNAP-associated factor 1; KIAA0857; Phosphoprotein pp75; pp75; RAB11 family interacting protein 5; RAB11 family interacting protein 5 (class I); rab11 family-interacting protein 5; Rab11fip5; Rab11-FIP5; rab11-interacting protein Rip11; Rip11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXF6 | |
| RAB11FIP5 | |
| Recombinant protein corresponding to Human RAB11FIP5. Recombinant protein control fragment (Product #RP-96081). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction