missing translation for 'onlineSavingsMsg'
Learn More

RAB11FIP5 Antibody, Novus Biologicals™

Product Code. 18326969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326969 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326969 Supplier Novus Biologicals Supplier No. H00026056B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RAB11FIP5 Polyclonal antibody specifically detects RAB11FIP5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB11FIP5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH35013
Gene Alias DKFZp434H018, Gaf-1, GAF1rab11-FIP5, Gamma-SNAP-associated factor 1, KIAA0857gaf-1, Phosphoprotein pp75, pp75, RAB11 family interacting protein 5 (class I), Rab11-FIP5, Rab11-interacting protein Rip11, RAB11RIP5, RIP11rab11 family-interacting protein 5
Host Species Mouse
Immunogen RAB11FIP5 (AAH35013, 1 a.a. - 652 a.a.) full-length human protein. MALVRGAEPAAGPSRWLPTHVQVTVLRARGLRGKSSGAGSTSDAYTVIQVGREKYSTSVVEKTHGCPEWREECSFELPPGALDGLLRAQEADAGPAPWAASSAAACELVLTTMHRSLIGVDKFLGQATVALDEVFGAGRAQHTQWYKLHSKPGKKEKERGEIEVTIQFTRNNLSAMFDLSMKDKPRSPFSKIRDKMKGKKKYDLESASAILPSSAIEDPDLGSLGKMGKAKGFFLRNKLRKSSLTQSNTSLGSDSTLSSASGSLAYQGPGAELLTRSPSRSSWLSTEGGRDSAQSPKLFTHKRTYSDEANQMRVAPPRALLDLQGHLDAASRSSLCVNGSHIYNEEPQGPVRHRSSISGSLPSSGSLQAVSSRFSEEGPRSTDDTWPRGSRSNSSSEAVLGQEELSAQAKVLAPGASHPGEEEGARLPEGKPVQVATPIVASSEAVAEKEGARKEERKPRMGLFHHHHQGLSRSELGRRSSLGEKGGPILGASPHHSSSGEGKAKSSWFGLREAKDPTQKPSPHPVKPLSAAPVEGSPDRKQSRSSLSIALSSGLEKLKTVTSGSIQPVTQAPQAGQMVDTKRLKDSAVLDQSAKYYNLTHDELISLLLQRERELSQRDEHVQELESYIDRLLVRIMETSPTLLQIPPGPPK
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26056
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.