missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11FIP4 Antibody (OTI13B5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73764H
This item is not returnable.
View return policy
Description
RAB11FIP4 Monoclonal specifically detects RAB11FIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RAB11FIP4 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| RAB11FIP4 | |
| Recombinant protein fragment corresponding to amino acids 91-177 of human RAB11FIP4 (NP_116321) produced in E.coli. | |
| 0.1 mL | |
| 84440.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI13B5 | |
| Western Blot, Immunohistochemistry | |
| ARFO2, Arfophilin-2, FIP4-Rab11, FLJ00131, KIAA1821arfophilin-2, MGC11316, MGC126566, RAB11 family interacting protein 4 (class II), rab11 family-interacting protein 4, Rab11-FIP4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction