missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB11FIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558078
This item is not returnable.
View return policy
Description
Immunogen sequence: HMPDANSEFS SGEIQMKSKP KKPFLLGPQR LSSAHSMSDL SGSHMSSEKL KAGTIGQTHL LGHQLDSFGT VPESGSLKSP HRRTLSFDTS KMN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.
Specifications
| RAB11FIP2 | |
| Polyclonal | |
| Unconjugated | |
| RAB11FIP2 | |
| 4930470G04Rik; A830046J09Rik; AW558126; KIAA0941; nRip11; RAB11 family interacting protein 2; RAB11 family interacting protein 2 (class I); rab11 family-interacting protein 2; rab11-family interacting protein 2; Rab11fip2; Rab11-FIP2; RAB11-FIP2 long isoform; RGD1308538 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L804 | |
| RAB11FIP2 | |
| Recombinant protein corresponding to Human RAB11FIP2. Recombinant protein control fragment (Product #RP-97030). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction