missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB11FIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550102
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total RAB11FIP2 protein.
RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.
Specifications
| RAB11FIP2 | |
| Polyclonal | |
| Unconjugated | |
| RAB11FIP2 | |
| 4930470G04Rik; A830046J09Rik; AW558126; KIAA0941; nRip11; RAB11 family interacting protein 2; RAB11 family interacting protein 2 (class I); rab11 family-interacting protein 2; rab11-family interacting protein 2; Rab11fip2; Rab11-FIP2; RAB11-FIP2 long isoform; RGD1308538 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22841, 74998 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| G3XA57, Q7L804 | |
| RAB11FIP2 | |
| Synthesized peptide derived from internal of human RAB11FIP2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction