missing translation for 'onlineSavingsMsg'
Learn More

RAB11B Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 67780-1-IG-20UL

 View more versions of this product

Product Code. 30236816

  • ONLINE EXCLUSIVE PRICE
    £135.00
    £119.00 / 20µL
    Save £16.00
    11% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RAB11B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB11B. This antibody recognizes human, mouse, rat antigen. The RAB11B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

RAB11B
Monoclonal
1000 μg/mL
H-YPT3; MGC133246
Recombinant Protein
20 μL
Primary
Human, Mouse, Rat
24 kDa
Liquid
Western Blot, ELISA
1G7D6
Unconjugated
Mouse
Protein G purification
RUO
9230
Store at -20°C. Stable for one year after shipment.
Monoclonal Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.