Learn More
Invitrogen™ RAB11A Recombinant Rabbit Monoclonal Antibody (3H18L5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 700184
Description
This antibody is predicted to react with bovine, chicken, equine, mouse, primate, rat, Xenopus and zebrafish based on sequence homology. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Rab11A is a member of the YPT-1 subfamily of Ras-related GTP-binding proteins. Rab11A mRNA has been detected in a variety of tissues including brain, testis, spleen, heart, and gastrointestinal mucosa. In the gastric fundus the Rab11A protein is expressed in parietal cell tubulovesicles membranes. Expression of the Rab11A protein appears to occur in discrete epithelial cell populations where it localizes to apical vesicular populations. Recent studies have shown Rab11A is essential for resecretion of alpha-synuclein out of neuronal cells via endosome recycling and is required for exocytosis of discoidal/fusiform vesicles of bladder umbrella cells. Rab11A is also implicated in biogenesis of Birbeck granules and endosomal activation of the PI3K/AKT signaling pathway via G beta gamma trafficking.
Specifications
| RAB11A | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P62491, P62492, P62494 | |
| RAB11A | |
| A peptide corresponding to amino acids 179-194 of P62491. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 3H18L5 | |
| Unconjugated | |
| RAB11A | |
| 24KG; RAB 11A, member oncogene family; RAB11; rab-11; rab11 GTP-binding protein; Rab11a; RAB11a, member RAS oncogene family; ras-related protein Rab-11A; tubulovesicle-associated protein; YL8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 53869, 81830, 8766 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.