missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab11A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3612-50ug
This item is not returnable.
View return policy
Description
Rab11A Polyclonal specifically detects Rab11A in Human, Porcine, Canine, Primate samples. It is validated for Western Blot.
Specifications
| Rab11A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RAB11A | |
| Synthetic peptide corresponding to residues M(1) G T R D D E Y D Y L F K V V L I C(17) of human Rab11. | |
| 50 μg | |
| Cancer, Tumor Suppressors | |
| 8766 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 6.6 μg/mL | |
| MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB120-3612 detects Rab 11 from human and non-human primate samples. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction