missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579901
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell, human Hela whole cell, human A549 whole cell, rat brain tissue, rat C6 whole cell, mouse brain tissue, mouse NIH/3T3 whole cell.
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.
Specifications
| RAB10 | |
| Polyclonal | |
| Unconjugated | |
| RAB10 | |
| Ac1075; AW107754; GTP-binding protein RAB10; Rab10; RAB10, member RAS oncogene family; ras-related GTP-binding protein; ras-related protein rab10; ras-related protein Rab-10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10890, 19325, 50993 | |
| -20°C | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P35281, P61026, P61027 | |
| RAB10 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human RAB10 (153-185aa AKANINIEKAFLTLAEDILRKTPVKEPNSENVD). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction