missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB10 Antibody (PSH03-24), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32882
This item is not returnable.
View return policy
Description
RAB10 Monoclonal antibody specifically detects RAB10 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| RAB10 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| GTP-binding protein RAB10, RAB10, member RAS oncogene family, ras-related GTP-binding protein, ras-related protein Rab-10 | |
| Synthetic peptide within human RAB10 aa 101-150/ 200. (Uniprot: P61026) | |
| 100 μL | |
| Signal Transduction | |
| 10890 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| PSH03-24 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction