missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ RA Lamb Five-Slide Mailer, holds 5 slides

Product Code. 12688556 Shop All Epredia Products
Click to view available options
Capacity:
5
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12688556

Brand: Epredia™ E6/3

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Hold five 76 x 26mm microscope slides with the Epredia™ RA Lamb 5-Slide Mailer, made of molded polypropylene.

  • Molded polypropylene construction
  • Attached snap-on lid and grooves
TRUSTED_SUSTAINABILITY

Specifications

Type RA Lamb Five-Slide Mailer
Dimensions (LxWxH) 76 mm x 26 mm
Capacity 5
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.