Learn More
Epredia™ RA Lamb Coverslip Staining Rack
Shop All Epredia Products
Click to view available options
Holds:
Slides
Unit Size:
Each
Description
Features
- Stainless steel construction adds durability
- Slot spacing permits good drainage and reagent exchange
Includes
- Spring-type detachable handle
Specifications
Specifications
| Product Type | Coverslip Staining Rack |
| Holds | Slides |
| Material | Stainless Steel |
| Positions | 25 |
| Length (Metric) Slide | 22 mm |
| Width (Metric) Slide | 22 mm |