missing translation for 'onlineSavingsMsg'
Learn More

R5C3, Thermo Scientific™

Product Code. 15447009
Change view
Click to view available options
Quantity:
10 mg
Packaging:
Plastic bottle
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15447009 10 mg Plastic bottle 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447009 Supplier Thermo Scientific Alfa Aesar Supplier No. J65639.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 753504-14-2
Packaging Plastic bottle
Molecular Formula C33H28N2O6
MDL Number MFCD08705330
Quantity 10 mg
Synonym 1-benzyloxy carbonyl-5-methyl-n-2-phenoxybenzoyl tryptophan, 5-methyl-n-2-phenoxybenzoyl-1-phenylmethoxy carbonyl-d/l-tryptophan, 3-5-methyl-1-phenylmethoxycarbonyl indol-3-yl-2-2-phenoxy benzoyl amino propanoic acid
InChI Key FHNQOOZTZWBWQO-UHFFFAOYSA-N
SMILES CC1=CC2=C(C=C1)N(C=C2CC(C(=O)O)NC(=O)C3=CC=CC=C3OC4=CC=CC=C4)C(=O)OCC5=CC=CC=C5
IUPAC Name 3-(5-methyl-1-phenylmethoxycarbonylindol-3-yl)-2-[(2-phenoxybenzoyl)amino]propanoic acid
Molecular Weight (g/mol) 548.595
PubChem CID 16079003
Formula Weight 548.58
Physical Form Powder
Chemical Name or Material R5C3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.