missing translation for 'onlineSavingsMsg'
Learn More

R406 free base, MedChemExpress

Product Code. 16494398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494398 10 mg 10mg
16490717 50 mg 50mg
16455296 100 mg 100mg
16407127 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494398 Supplier Medchem Express Supplier No. HY11108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R406 free base is an orally available and competitive Syk/FLT3 inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. R406 free base reduces immune complex-mediated inflammation. R406 free base also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

R406 free base is an orally available and competitive Syk/FLT3 inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. R406 free base reduces immune complex-mediated inflammation. R406 free base also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

Specifications

Chemical Name or Material R406 free base
CAS 841290-80-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H23FN6O5
Quantity 10 mg
Solubility Information DMSO : ≥ 10 mg/mL (21.26 mM)
SMILES COC1=CC(NC2=NC(NC3=NC(N4)=C(C=C3)OC(C)(C)C4=O)=C(C=N2)F)=CC(OC)=C1OC
Molecular Weight (g/mol) 470.45
Formula Weight 470.45
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.