missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Specifications
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| For Use With (Application) | Protein Sequencing & Peptide Synthesis |
| Product Type | Protein Sequencing Reagents |
| Quantity | 1 Ea. |
| Reagent Type | N-terminal Sequencing Reagent |
| Shipping Condition | Room Temperature |
| Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?