missing translation for 'onlineSavingsMsg'
Learn More

Curcumol, MedChemExpress

Product Code. 16418868
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16418868

Brand: Medchem Express HYN0104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Curcumol ((-)-Curcumol), a bioactive sesquiterpenoid, possesses numerous pharmacological activities like anticancer, antimicrobial, antifungal, antiviral, and antiinflammatory. Curcumol is a potent inducer of apoptosis in numerous cancer cells via targeting key signaling pathways as MAPK/ERK, PI3K/Akt and NF-κB which are generally deregulated in several cancers.

Curcumol ((-)-Curcumol), a bioactive sesquiterpenoid, possesses numerous pharmacological activities like anticancer, antimicrobial, antifungal, antiviral, and antiinflammatory. Curcumol is a potent inducer of apoptosis in numerous cancer cells via targeting key signaling pathways as MAPK/ERK, PI3K/Akt and NF-κB which are generally deregulated in several cancers.

Specifications

Chemical Name or Material Curcumol
Physical Form Solid
CAS 4871-97-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H24O2
For Use With (Application) Cancer-Kinase/protease
Synonym (-)-Curcumol
Solubility Information DMSO : ≥ 100 mg/mL (423.10 mM)
SMILES O[C@@]1(C2)[C@H](C(C)C)C[C@]3(O1)[C@@H](C)CC[C@@]3([H])C2=C
Molecular Weight (g/mol) 236.35
Formula Weight 236.35
Percent Purity 98.62%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.