missing translation for 'onlineSavingsMsg'
Learn More

R121919, MedChemExpress

Product Code. p-200096582
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492107

Brand: Medchem Express HY14127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R121919 (NBI30775) is a potent small-molecule CRF1 receptor antagonist with a Ki of 2 to 5 nM for the CRF1 receptor and over 1000-fold weaker activity at the CRF2 receptor, CRF-binding protein, or 70 other receptor types.

R121919 (NBI30775) is a potent small-molecule CRF1 receptor antagonist with a Ki of 2 to 5 nM for the CRF1 receptor and over 1000-fold weaker activity at the CRF2 receptor, CRF-binding protein, or 70 other receptor types.

Specifications

Chemical Name or Material R121919
CAS 195055-03-9
Color White
Purity Grade Notes Research
Molecular Formula C22H32N6
Quantity 50 mg
Synonym NBI30775
Solubility Information Ethanol : 100 mg/mL (262.79 mM; Need ultrasonic) ∣DMSO : 6.2 mg/mL (16.29 mM; Need ultrasonic)
SMILES CCCN(C1=CC(C)=NC2=C(C3=CN=C(N(C)C)C=C3C)C(C)=NN21)CCC
Molecular Weight (g/mol) 380.53
Formula Weight 380.53
Percent Purity 99.45%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.