missing translation for 'onlineSavingsMsg'
Learn More

(R)-Usnic acid Sodium, TRC

Product Code. 30456944
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456944 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456944 Supplier Toronto Research Chemicals Supplier No. TRCU850155100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-Usnic acid Sodium
CAS 2796975-82-9
Molecular Formula C18 H15 Na O7
Quantity 100 mg
Synonym sodium (R)-2,6-diacetyl-7,9-dihydroxy-8,9b-dimethyl-1-oxo-1,9b-dihydrodibenzo[b,d]furan-3-olate
InChI Formula InChI=1S/C18H16O7.Na/c1-6-14(22)12(8(3)20)16-13(15(6)23)18(4)10(25-16)5-9(21)11(7(2)19)17(18)24;/h5,21-23H,1-4H3;/q;+1/p-1/t18-;/m0./s1
SMILES CC(=O)C1=C(O[Na])C=C2Oc3c(C(=O)C)c(O)c(C)c(O)c3[C@@]2(C)C1=O
IUPAC Name [(9bR)-2,6-diacetyl-7,9-dihydroxy-8,9b-dimethyl-1-oxodibenzofuran-3-yl]oxysodium
Molecular Weight (g/mol) 366.297
Formula Weight 366.072
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.