missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Tolvaptan, (R)-(+)-Tolvaptan, TRC

Product Code. 18092330
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18092330 1 mg 1mg
17919849 2.5 mg 2.5mg
18082920 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092330 Supplier Toronto Research Chemicals Supplier No. TRCT5366401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C26 H25 Cl N2 O3
Molecular Weight (g/mol) 448.94
Synonym N-[4-[[(5R)-7-Chloro-2,3,4,5-tetrahydro-5-hydroxy-1H-1-benzazepin-1-yl]carbonyl]-3-methylphenyl]-2-methylbenzamide, (R)-(+)-OPC 41061
IUPAC Name N-[4-[(5S)-7-chloro-5-hydroxy-2,3,4,5-tetrahydro-1-benzazepine-1-carbonyl]-3-methylphenyl]-2-methylbenzamide
SMILES Cc1ccccc1C(=O)Nc2ccc(C(=O)N3CCC[C@H](O)c4cc(Cl)ccc34)c(C)c2

Specifications

CAS 331947-66-1
Quantity 1 mg
InChI Formula InChI=1S/C26H25ClN2O3/c1-16-6-3-4-7-20(16)25(31)28-19-10-11-21(17(2)14-19)26(32)29-13-5-8-24(30)22-15-18(27)9-12-23(22)29/h3-4,6-7,9-12,14-15,24,30H,5,8,13H2,1-2H3,(H,28,31)/t24-/m0/s1
Formula Weight 448.1554
Chemical Name or Material (R)-(+)-Tolvaptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.