missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%

Product Code. 11378706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378706 25 g 25g
11358706 1 g 1g
11368706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378706 Supplier Thermo Scientific Alfa Aesar Supplier No. L19410.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87392-05-0 | C5H8O3 | 116.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87392-05-0
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00211271
InChI Key UJJLJRQIPMGXEZ-SCSAIBSYSA-N
Synonym r-+-2-tetrahydrofuroic acid, r-+-tetrahydro-2-furoic acid, r-tetrahydrofuran-2-carboxylic acid, r-+-tetrahydrofuran-2-carboxylic acid, r-2-tetrahydrofuroic acid, 2-furancarboxylic acid, tetrahydro-, 2r, 2r-oxolane-2-carboxylic acid, unii-7396hfq44j, r-tetrahydro-2-furoic acid, d-tetrahydro-furan-2-carboxylic acid
PubChem CID 2734707
IUPAC Name (2R)-oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

Density 1.213
Quantity 25 g
Refractive Index 1.46
UN Number UN3265
Beilstein 4658739
Boiling Point 244°C
Formula Weight 116.12
Percent Purity ≥98%
Assay ee 98%
Chemical Name or Material (R)-(+)-Tetrahydro-2-furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.