missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-tert-Leucinol, 98%

Product Code. 10775832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10775832 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10775832 Supplier Thermo Scientific Acros Supplier No. 436671000
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 112245-09-7 | C6H15NO | 117.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 112245-09-7
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C6H15NO
Packaging Glass Bottle
MDL Number MFCD01076574
Specific Rotation -34° to -40° (20°C, 589nm) (c=0.7, ethanol)
Synonym r-tert-leucinol, d-tert-leucinol, 2r-2-amino-3,3-dimethylbutan-1-ol, r-2-amino-3,3-dimethylbutan-1-ol, r-2-amino-3,3-dimethyl-1-butanol, 2r-2-amino-3,3-dimethyl-1-butanol, r---tert-leucinol, ?-tert-leucinol, r-?-tert-leucinol
InChI Key JBULSURVMXPBNA-MLHKIVSYNA-N
Density 0.9g/mL
SMILES CC(C)(C)[C@@H](N)CO
IUPAC Name (2R)-2-amino-3,3-dimethylbutan-1-ol
Molecular Weight (g/mol) 117.19
Quantity 100 mg
PubChem CID 6932847
Formula Weight 117.19
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material (R)-(-)-tert-Leucinol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-tert-Leucinol, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.