missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-tert-Butylsulfinamide, 98%

Product Code. 10753722 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10753722 5 g Glass bottle 5g
10530359 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753722 Supplier Thermo Scientific Acros Supplier No. 391100050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 196929-78-9 | C4H11NOS | 121.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196929-78-9
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD05861479
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide
PubChem CID 10964479
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 99.0°C to 102.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSONH2
Quantity 5 g
Merck Index 15,3607
Specific Rotation 3.0° to 5.0° (20°C, 589nm) (c=1, chloroform)
Formula Weight 121.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-(+)-tert-Butylsulfinamide
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-tert-Butylsulfinamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.