missing translation for 'onlineSavingsMsg'
Learn More

(R)-tert-Butyl 3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]piperidine-1-carboxylate, Mikromol

Product Code. 30463580
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30463580 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463580 Supplier Mikromol Supplier No. MM3746.020025
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(R)-tert-Butyl 3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]piperidine-1-carboxylate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-Ibrutinib ( tert-Butyl-1-carboxyl Piperidine analog of Ibrutinib)
CAS 1022150-11-3
Analyte or Component Names N-Boc-Ibrutinib ( tert-Butyl-1-carboxyl Piperidine analog of Ibrutinib)
Shipping Condition Room Temperature
Molecular Formula C27H30N6O3
Certifications/Compliance ISO 9001
Synonym 1-Piperidinecarboxylic acid, 3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-, 1,1-dimethylethyl ester, (3R)- (ACI), 1,1-Dimethylethyl (3R)-3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinecarboxylate (ACI), (R)-3-[4-Amino-3-(4-phenoxy-phenyl)-pyrazolo[3,4-d]pyrimidin-1-yl]-piperidine-1-carboxylic acid tert-butyl ester, (R)-tert-Butyl 3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]piperidine-1-carboxylate
InChI Formula InChI=1S/C27H30N6O3/c1-27(2,3)36-26(34)32-15-7-8-19(16-32)33-25-22(24(28)29-17-30-25)23(31-33)18-11-13-21(14-12-18)35-20-9-5-4-6-10-20/h4-6,9-14,17,19H,7-8,15-16H2,1-3H3,(H2,28,29,30)/t19-/m1/s1
Solution Type Neat
SMILES CC(C)(C)OC(=O)N1CCC[C@H](C1)n2nc(c3ccc(Oc4ccccc4)cc3)c5c(N)ncnc25
IUPAC Name tert-butyl (3R)-3-[4-amino-3-(4-phenoxyphenyl)pyrazolo[3,4-d]pyrimidin-1-yl]piperidine-1-carboxylate
Molecular Weight (g/mol) 486.57
Quantity 25 mg
Formula Weight 486.2379 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.