missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Styrene Oxide, TRC

Product Code. 30414787
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414787 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414787 Supplier Toronto Research Chemicals Supplier No. TRCS6878001G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(+)-Styrene Oxide
CAS 20780-53-4
Molecular Formula C8 H8 O
Quantity 1 g
Synonym (R)-Phenyloxirane, 2-Phenyl-oxirane, (R)-(+)-(Epoxyethyl)benzene, (2R)-2-Phenyloxirane, (R)-Epoxystyrene, (R)-Phenylethylene Oxide
InChI Formula InChI=1S/C8H8O/c1-2-4-7(5-3-1)8-6-9-8/h1-5,8H,6H2/t8-/m0/s1
SMILES C1O[C@@H]1c2ccccc2
IUPAC Name (2R)-2-phenyloxirane
Molecular Weight (g/mol) 120.148
Formula Weight 120.058
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.