missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Styrene oxide, 98%

Product Code. 11444767
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444767 1 g 1g
11454767 5 g 5g
11464767 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444767 Supplier Thermo Scientific Alfa Aesar Supplier No. B21245.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 20780-53-4 | C8H8O | 120.15 g/mol

It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20780-53-4
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00066210
InChI Key AWMVMTVKBNGEAK-QMMMGPOBSA-N
Synonym r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r
PubChem CID 114705
ChEBI CHEBI:45389
IUPAC Name (2R)-2-phenyloxirane
SMILES C1O[C@@H]1C1=CC=CC=C1

Specifications

Boiling Point 194°C
Flash Point 74°C (165°F)
Quantity 1 g
UN Number UN2810
Beilstein 1984
Optical Rotation +32° (Neat)
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material (R)-(+)-Styrene oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.