missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse R-Spondin 4 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | R-Spondin 4 |
---|---|
Concentration | LYOPH |
Dilution | Neutralization 5.0-20 ug/mL |
Applications | Neutralization |
Classification | Polyclonal |
Description
R-Spondin 4 Polyclonal specifically detects R-Spondin 4 in Mouse samples. It is validated for Neutralization.Specifications
R-Spondin 4 | |
Neutralization 5.0-20 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
C20orf182, chromosome 20 open reading frame 182, Cristin 4, CRISTIN4, dJ824F16.3, FLJ16018, hRspo4, Roof plate-specific spondin-4, RSPO4, RSpondin 4, R-spondin family, member 4, R-spondin-4 | |
RSPO4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Neutralization | |
Unconjugated | |
RUO | |
Q8BJ73 | |
343637 | |
E. coli-derived recombinant mouse R-Spondin 4 Tyr21-Pro197 Accession # Q8BJ73 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse R-Spondin 4 in direct ELISAs. |
For Research Use Only