missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 4 Antibody (435203) [Biotin], Novus Biologicals™

Product Code. 30731503 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731503 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731503 Supplier Novus Biologicals Supplier No. FAB4106B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

R-Spondin 4 Monoclonal antibody specifically detects R-Spondin 4 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 4
Applications Western Blot
Classification Monoclonal
Clone 435203
Conjugate Biotin
Formulation PBS
Gene Alias C20orf182, chromosome 20 open reading frame 182, CRISTIN4, dJ824F16.3, FLJ16018, hRspo4, Roof plate-specific spondin-4, R-spondin family, member 4, R-spondin-4
Host Species Rat
Immunogen E. coli-derived recombinant mouse R-Spondin 4, Tyr21-Pro197, Accession # Q8BJ73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343637
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.