missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 3 Antibody (400403) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30730938 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30730938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730938 Supplier Novus Biologicals Supplier No. FAB41201J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

R-Spondin 3 Monoclonal antibody specifically detects R-Spondin 3 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 3
Applications Western Blot
Classification Monoclonal
Clone 400403
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Host Species Rat
Immunogen E. coli-derived recombinant mouse R-Spondin 3, Ser21-Gly209 (Gln108Lys), Accession # BAC36296
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84870
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.