missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ R-Spondin 2 Antibody (CL14321), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26851-25ul
This item is not returnable.
View return policy
Description
R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Human,Mouse samples. It is validated for Immunohistochemistry
Specifications
| R-Spondin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:200 - 1:500 | |
| CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2 | |
| This antibody was developed against a Synthetic Peptide of Q6UXX9. Actual immunogen sequence is proprietary. | |
| 25 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry | |
| CL14321 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 340419 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction