missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 2 Antibody (879712R) [DyLight 488], Novus Biologicals™

Product Code. 30731284 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731284 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731284 Supplier Novus Biologicals Supplier No. FAB32662K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 2
Applications ELISA
Classification Monoclonal
Clone 879712R
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human R-Spondin 2, Met1-Gly205, Accession # Q6UXX9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340419
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.