missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | R-Spondin 2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 879712R |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2 |
| Host Species | Rat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human R-Spondin 2, Met1-Gly205, Accession # Q6UXX9 |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?