missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 2 Antibody (773029) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30733756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733756 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733756 Supplier Novus Biologicals Supplier No. FAB32661JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Mouse samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 2
Applications Neutralization
Classification Monoclonal
Clone 773029
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse R-Spondin 2, Met1-Gly205, Accession # Q8BFU0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340419
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.