missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R-Spondin 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77354MFV610
This item is not returnable.
View return policy
Description
R-Spondin 1 Polyclonal antibody specifically detects R-Spondin 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| R-Spondin 1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1 | |
| Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human R-Spondin 1. The immunogen is located within the first 50 amino acids of R-Spondin 1. Amino Acid Sequence: RGIKGKRQRRISAEGS | |
| 0.1 mL | |
| Signal Transduction | |
| 284654 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction