missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 1 Antibody (852010) [PerCP], Novus Biologicals™

Product Code. 30737542 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737542 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737542 Supplier Novus Biologicals Supplier No. FAB3474C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

R-Spondin 1 Monoclonal antibody specifically detects R-Spondin 1 in Mouse samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 1
Applications Neutralization
Classification Monoclonal
Clone 852010
Conjugate PerCP
Formulation PBS
Gene Alias CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse R-Spondin 1, Met1-Gln265, Accession # Q9Z132
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 284654
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.