missing translation for 'onlineSavingsMsg'
Learn More

(R,S,R)-2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)-imidazole, TRC

Product Code. 30133448
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133448 500 mg 500mg
30010080 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133448 Supplier Toronto Research Chemicals Supplier No. TRCA188490500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94944-70-4
Molecular Formula C9H14N2O5
Molecular Weight (g/mol) 230.22
Synonym 1-[5-[(1R,2S,3R)-1,2,3,4-Tetrahydroxybutyl]-1H-imidazol-2-yl]-ethanone, [1R-(1R*,2S*,3R*)]-1-[4-(1,2,3,4-Tetrahydroxybutyl)-1H-imidazol-2-yl]-ethanone
IUPAC Name 1-[4-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]-1H-imidazol-2-yl]ethanone
SMILES CC(C1=NC([C@H]([C@@H]([C@@H](CO)O)O)O)=CN1)=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H14N2O5/c1-4(13)9-10-2-5(11-9)7(15)8(16)6(14)3-12/h2,6-8,12,14-16H,3H2,1H3,(H,10,11)/t6-,7-,8-/m1/s1
Formula Weight 230.0903
Chemical Name or Material (R,S,R)-2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.