missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-Palonosetron Hydrochloride (90%), TRC

Product Code. 30418419
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418419 25 mg
30419337 10 mg
30414932 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418419 Supplier Toronto Research Chemicals Supplier No. TRCP16579525MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135755-51-0
Molecular Formula C19 H24 N2 O . Cl H
Molecular Weight (g/mol) 332.87
Synonym 1H-Benz[de]isoquinolin-1-one, 2-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-, hydrochloride (1:1), (3aR)-, 1H-Benz[de]isoquinolin-1-one, 2-(1-azabicyclo[2.2.2]oct-3-yl)-2,3,3a,4,5,6-hexahydro-, monohydrochloride, [S-(R*,S*)]-, 1H-Benz[de]isoquinolin-1-one, 2-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-, monohydrochloride, (3aR)- (9CI), (3aR)-2-(3S)-1-Azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-1H-benz[de]isoquinolin-1-one hydrochloride (1:1), RS 25233-198
IUPAC Name (3aR)-2-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one;hydrochloride
SMILES Cl.O=C1N(C[C@@H]2CCCc3cccc1c23)[C@@H]4CN5CCC4CC5

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17+;/m0./s1
Formula Weight 332.1655
Chemical Name or Material (R,S)-Palonosetron Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.